missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ AKR1B1 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human AKR1B1 (P15121) (Met 1-Phe 316) fused to a polyhistidine tag at the N-terminus

Manufacturer:  Life Technologies LS11294H07E250

 View more versions of this product

Catalog No. 11294H07EL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; AKR1B1 Recombinant Human Protein, N-His tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Aldo-Keto Reductase family 1 member B1 (aldose reductase) (AKR1B1), recombinant human protein is supplied frozen. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: PBS, 20% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
231
PAGE
5 x 50μg
Full Length/Recombinant
Aldo-Keto Reductase Family 1, Member B1 (Aldose Reductase)
Assay Standard
E. coli
Frozen
37.9kDa
≥85% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
ADR, ALDR1, ALR2, AR, mgC1804
AKR1B1
Other Proteins
His Tag