missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AIM2 Recombinant Human Protein, N-GST Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human AIM2 (NP_004824.1) (Met 1-Thr 343) fused to the GST tag at the N-terminus

$564.00 - $564.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AIM2, RP11-520H16.3, PYHIN4
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11654H09B50 View Documents Life Technologies
LS11654H09B50
50μg Each for $564.00
 
Description

Description

Absent In Melanoma 2 (AIM2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells (Karey, K.P. et al. (1988) Cancer Research 48:4083); the ED50 for this effect is typically 4-7μg/mL
  • Formulation: Lyophilized in 50mM Tris, 1M NaCl, 0.5mM PMSF, 5mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
AIM2, RP11-520H16.3, PYHIN4

For Research Use Only. Not for use in diagnostic procedures.