Please login to your online account to display your discounted pricing

Sino Biological™ AIM2 Recombinant Human Protein, N-GST tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human AIM2 (NP_004824.1) (Met 1-Thr 343) fused to the GST tag at the N-terminus

Manufacturer: life technologies  LS11654H09B250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11654H09BL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 65.2kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 50μg
Protein Subtype NFκB Pathway Proteins
Common Name Absent in melanoma 2
Protein Family Other Proteins
Expression System Insect
Format Lyophilized
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Endotoxin Concentration <1 EU/μg
Gene Alias AIM2, RP11-520H16.3, PYHIN4
Gene Symbol AIM2
Protein Tag GST Tag
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Absent In Melanoma 2 (AIM2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells (Karey, K.P. et al. (1988) Cancer Research 48:4083); the ED50 for this effect is typically 4-7μg/mL
  • Formulation: Lyophilized in 50mM Tris, 1M NaCl, 0.5mM PMSF, 5mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.