missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AGER Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human AGER isoform 1 (NP_001127.1) (Met 1-Ala 344) fused to the Fc region of human IgG1 at the C-terminus

$102.00 - $102.00

Specifications

Purity >70% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias DAMA-358M23.4, mgC22357, RAGE
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11629H02H5 View Documents Life Technologies
LS11629H02H5
5μg Each for $102.00
 
Description

Description

Advanced glycosylation end product-specific receptor (AGER), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human S100A12 at 2μg/mL (100μL/well) can bind recombinant human AGER with a linear range of 0.032-20μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

>70% by SDS-PAGE
<1 EU/μg
Lyophilized
DAMA-358M23.4, mgC22357, RAGE

For Research Use Only. Not for use in diagnostic procedures.