Please login to your online account to display your discounted pricing

Sino Biological™ AGER Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human AGER isoform 1 (NP_001127.1) (Met 1-Ala 344) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS11629H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11629H02H50

  • / Each
 

Description & Specifications

Specifications

Purity >70% by SDS-PAGE
Common Name Advanced glycosylation end product-specific receptor
Protein Family Other Proteins
Protein Tag IgG1 Fc Tag
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Protein Subtype Other Proteins
Expression System Mammalian
Gene Alias DAMA-358M23.4, mgC22357, RAGE
Gene Symbol AGER
Validated Application Assay Standard
Quantity 50μg
Endotoxin Concentration <1 EU/μg
Molecular Weight 61.1kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.

Advanced glycosylation end product-specific receptor (AGER), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human S100A12 at 2μg/mL (100μL/well) can bind recombinant human AGER with a linear range of 0.032-20μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.