missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AGER Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human AGER isoform 1 (NP_001127.1) (Met 1-Ala 344) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer:  Invitrogen LS11629H02H5

 View more versions of this product

Catalog No. 11629H02H5



Description

Description

Advanced glycosylation end product-specific receptor (AGER), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human S100A12 at 2μg/mL (100μL/well) can bind recombinant human AGER with a linear range of 0.032-20μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
61.1kDa
5μg
<1 EU/μg
Advanced glycosylation end product-specific receptor
IgG1 Fc Tag
Other Proteins
Assay Standard
Lyophilized
>70% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
DAMA-358M23.4, mgC22357, RAGE
AGER
SDS
Documents
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.