missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ACVR1B (ALK4) Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ACVR1B (NP_004293.1) extracellular domain (Met 1-Glu 126) fused to C-terminal His-tagged Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10583H03H50

 View more versions of this product

Catalog No. 10583H03H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; ACVR1B (ALK4) Recombinant Human Protein (without catalytic activity domain), hIgG1-Fc His tag; 50ug

Activin A Receptor, Type Ib (ACVR1B), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TDGF1 at 2μg/mL (100μL/well) can bind human ACVR1B with a linear range of 0.0068-0.16μg/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Growth Factors & Receptors