missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ACPP Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ACPP (NP_001127666.1) (Met 1-Lys 382) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10959H08H25

 View more versions of this product

Catalog No. 10959H08H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; ACPP Recombinant Human Protein, His tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Acid phosphatase, prostate (ACPP), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave a substrate, pNitrophenyl phosphate (pNPP), the specific activity is >100nmol/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
42kDa
5 x 5μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Acid phosphatase, prostate
Assay Standard
His Tag
Lyophilized
≥90% by SDS-PAGE
Human
Other Proteins
ACP-3, ACP3, PAP
ACPP
Mammalian