Please login to your online account to display your discounted pricing

Sino Biological™ ACP1 Recombinant Human Protein, GST Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding human ACP1 (AAI06012.1) (Met 1-His 158) fused to a GST tag at the N-terminus

$111.00 - $1,124.00

Specifications

Purity ≥85% by SDS-PAGE
Format Lyophilized
Gene Alias HAAP, mgC111030, mgC3499
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10957H09E25

DFS Item
 
life technologies
LS10957H09E25
5 x 5μg Each for $241.00

10957H09E5

DFS Item
 
life technologies
LS10957H09E5
5μg Each for $111.00

10957H09E50

DFS Item
 
life technologies
LS10957H09E50
50μg Each for $372.00

10957H09EL

DFS Item
 
life technologies
LS10957H09E250
5 x 50μg Each for $1,124.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Format Lyophilized
Gene Alias HAAP, mgC111030, mgC3499

Acid phosphatase 1, soluble (ACP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave a substrate, pNitrophenyl phosphate (pNPP), the specific activity is >65nmol/min./μg
  • Formulation: Lyophilized in 50mM Tris, 150mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.