Please login to your online account to display your discounted pricing

Sino Biological™ ACP1 Recombinant Human Protein, GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding human ACP1 (AAI06012.1) (Met 1-His 158) fused to a GST tag at the N-terminus

Manufacturer: life technologies  LS10957H09E25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10957H09E25

  • / Each
 

Description & Specifications

Specifications

Source Human
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 5 x 5μg
Molecular Weight 44.3kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias HAAP, mgC111030, mgC3499
Protein Family Other Proteins
Expression System E. coli
Protein Tag GST Tag
Format Lyophilized
Protein Subtype Other Proteins
Gene Symbol ACP1
Common Name Acid phosphatase 1, soluble
Validated Application Assay Standard
Purity ≥85% by SDS-PAGE

Acid phosphatase 1, soluble (ACP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave a substrate, pNitrophenyl phosphate (pNPP), the specific activity is >65nmol/min./μg
  • Formulation: Lyophilized in 50mM Tris, 150mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.