missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ACO2 Recombinant Human Protein, N-His.GST Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ACO2 (Q99798) (Gln 28-Gln 780) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

$257.00 - $257.00

Specifications

Purity >85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias ACONM, mgC20605, mgC33908
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10958H20B5 Life Technologies
LS10958H20B5
5μg Each for $257.00
Specifications

Description

Aconitase 2 mitochondrial (ACO2), recombinant human protein is supplied in frozen format. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, 0.3mM DTT, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>85% by SDS-PAGE
<1 EU/μg
ACONM, mgC20605, mgC33908
PAGE
Frozen
50