Please login to your online account to display your discounted pricing

Sino Biological™ ACO2 Recombinant Human Protein, N-His.GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human ACO2 (Q99798) (Gln 28-Gln 780) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Manufacturer: life technologies  LS10958H20B5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10958H20B5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 110kDa
Purification Method PAGE
Purity >85% by SDS-PAGE
Quantity 5μg
Source Human
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias ACONM, mgC20605, mgC33908
Common Name Aconitase 1, Mitochondrial
Gene Symbol ACO2
Protein Tag GST Tag/His Tag
Validated Application Assay Standard
Protein Family Mitochondrial Proteins
Expression System Insect
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen
Gene ID (Entrez) 50

Aconitase 2 mitochondrial (ACO2), recombinant human protein is supplied in frozen format. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, 0.3mM DTT, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.