missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ACLY Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ACLY (P53396) (Met 1-Met 1101) fused to a polyhistidine tag at the N-terminus

$147.00 - $147.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias ACL, ATPCL, CLATP
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11769H07B5 View Documents Life Technologies
LS11769H07B5
5μg Each for $147.00
 
Description

Description

ATP Citrate Lyase (ACLY), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
ACL, ATPCL, CLATP
PAGE
Frozen
47

For Research Use Only. Not for use in diagnostic procedures.