Please login to your online account to display your discounted pricing

Sino Biological™ ACLY Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human ACLY (P53396) (Met 1-Met 1101) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS11769H07B25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11769H07B25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 123kDa
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Common Name ATP Citrate Lyase
Protein Tag His Tag
Expression System Insect
Format Frozen
Purity ≥85% by SDS-PAGE
Quantity 5 x 5μg
Gene Alias ACL, ATPCL, CLATP
Gene Symbol ACLY
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purification Method PAGE
Source Human
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Family Other Proteins
Gene ID (Entrez) 47

ATP Citrate Lyase (ACLY), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.