missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ACHE Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse ACHE (NP_033729.1) (Met 1-Leu 614) fused to a polyhistidine tag at the C-terminus

$217.00 - $217.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias mE1a, mE1b, mE1c, mE1c-long, mE1d, mE1d', mE1e
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50543M08H5 View Documents Life Technologies
LS50543M08H5
5μg Each for $217.00
 
Description

Description

Acetylcholinesterase (ACHE), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave acetylthiocholine; the specific activity is >250nM/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
mE1a, mE1b, mE1c, mE1c-long, mE1d, mE1d', mE1e

For Research Use Only. Not for use in diagnostic procedures.