missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ACE2 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 740) of human ACE2 precursor (NP_068576.1) fused to Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10108H02H25

 View more versions of this product

Catalog No. 10108H02H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; ACE2 Recombinant Human Protein, hIgG1-Fc tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Angiotensin-Converting Enzyme 2 (ACE2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM Glycine, 10mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
59272
PAGE
5 x 5μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
Angiotensin I converting enzyme (peptidyl-dipeptidase A) 2
Assay Standard
Mammalian
Lyophilized
110.3kDa
≥90% by SDS-PAGE
Human
C-Terminal
Glycoproteins|Peptidase
ACE2, ACEH, DKFZp434A014
ACE2
Proteases, Peptidases & Inhibitors
IgG1 Fc Tag