missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

A2M Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human A2M (NP_000005.2) (Met 1-Ala 1474) fused to a polyhistidine tag at the C-terminus

$143.00 - $558.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias A2mp
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10952H08B5 View Documents Invitrogen
LS10952H08B5
5μg Each for $143.00
 
10952H08B50 View Documents Invitrogen
LS10952H08B50
50μg Each for $558.00
 
Description

Description

Alpha-2-Macroglobulin (A2M), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to trap trypsin; the trapped trypsin is no longer able to interact with protein substrates or inhibitors, but still able to cleave small peptide substrates or inhibitors, the IC50 value is <5nM
  • Formulation: 20mM Tris, 500mM NaCl, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Frozen
A2mp

For Research Use Only. Not for use in diagnostic procedures.