missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ A2M Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human A2M (NP_000005.2) (Met 1-Ala 1474) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10952H08B50

 View more versions of this product

Catalog No. 10952H08B50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; A2M Recombinant Human Protein, His tag; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Alpha-2-Macroglobulin (A2M), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to trap trypsin; the trapped trypsin is no longer able to interact with protein substrates or inhibitors, but still able to cleave small peptide substrates or inhibitors, the IC50 value is <5nM
  • Formulation: 20mM Tris, 500mM NaCl, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
164kDa
50μg
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
alpha-2-macroglobulin
Cell Structure/Organelle Proteins/Phosphatase Inhibitors
His Tag
Frozen
≥90% by SDS-PAGE
Human
Protease Inhibitors
A2mp
A2M
Insect