Epredia™ Single Cytofunnel™ with White Filter Cards
Hold sample volumes of up to 0.5 mL with the Epredia™ Single Cytofunnel™ with White Filter Cards.
$382.15 - $4,041.24
Specifications
Color | White |
---|---|
Disposable | Yes |
Type | Single Cytofunnel™, With Filter Cards and Caps |
Volume (Metric) | 0.1 to 0.5 mL |
Description
- Disposable sample chambers with white filter cards and caps
- Pre-attached filter card
Specifications
White | |
Yes | |
Single Cytofunnel™, With Filter Cards and Caps | |
0.1 to 0.5 mL |