missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sinapinic Acid MALDI Matrix, ProteoSpec™ Ricca Chemical

MALDI Matrices

Manufacturer:  Ricca Chemical Company RIRMB151035X10CM

 View more versions of this product

Catalog No. MB15103510C



Specifications

Specifications

ProteoSpec&trade Sinapinic Acid MALDI Matrix
Yellow
5 x 10mg
100
C11H12O5
Soluble in water
COC1=CC(=CC(=C1O)OC)C=CC(=O)O
224.212
CHEBI:15714
Solid
Molecular Biology
Glass Ampule
202°C
530-59-6
100
sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PCMORTLOPMLEFB-ONEGZZNKSA-N
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
637775
224.21
99.0% min.

Chemical Identifiers

530-59-6
224.212
sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
CHEBI:15714
COC1=CC(=CC(=C1O)OC)C=CC(=O)O
C11H12O5
PCMORTLOPMLEFB-ONEGZZNKSA-N
637775
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Recommended Storage : Refridgerated

Documents