Please login to your online account to display your discounted pricing

Simulated Gastric Fluid (without enzyme), J.T.Baker™

$659.98 - $738.34

Chemical Identifiers

Chemical Name or Material Simulated Gastric Fluid
CAS 7647-01-0
Name Note without enzyme
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

02004272

 
avantor performance
D02000
Each for $738.34

02004273

 
avantor performance
D02002
Each for $659.98
Description & Specifications

Specifications

Odor Acrid odor, sharp like hydrochloric acid
Color Colorless
Physical Form Liquid
CAS 7647-01-0
pH 0.4
Quantity 230.8mL, 12/Case; Poly Bottle
Solubility Information Completely Soluble
Name Note without enzyme
Density Estimated 1 (20°C)
Chemical Name or Material Simulated Gastric Fluid