Simport™ Scientific T100 BIOTUBE™ Tube Racks
Suitable for use in refrigerator, freezer or water bath, ideal for freezing and storing samples
Manufacturer:  Simport™ Scientific T1001B
 View more versions of this product
Catalog No. 05-500
Description
Description
Stands on four legs and can be removed from the base of the box and placed on a lab counter as a self-standing support
  • Can be placed in a refrigerator or freezer shelf for improved air circulation around tubes, or in a water bath to allow controlled warming of the tubes and their contents
  • Standard 96-well on center spacing of tubes
  • Color coding system using colored interchangeable plastic grids
  • Easy to read numbers and letters used on the box cover for sample identification
  • Also ideal for storing, freezing and transporting reagents and specimens
  • Availaible in red, blue, green and yellow
Specifications
Specifications
96
1.2mL tubes
96 Plain Individual Tubes
10/Cs.
T100 BIOTUBE Tube Rack
Blue
Tubes (1.2mL)
Polypropylene
Non-sterile