Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassettes with Hinged Covers
Disposable plastic Swingsette™ Tissue Processing/Embedding Cassettes hold tissue specimens during processing, embedding and storage.
Supplier: Simport™ Scientific M5154
Description
- Molded from a special high-density polymer, the cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
- Special hinge holds base and cover together, allowing cassettes to be opened and closed repeatedly
- Cover can be removed and re-inserted easily without danger of specimen loss
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
Specifications
Yes | |
Green | |
High Density Polymer | |
Tissue Cassette |