Please login to your online account to display your discounted pricing

Simport™ Scientific The UniRack™ Jr. Tube Racks

$43.08 - $43.36

Specifications

Autoclavable No
For Use With (Equipment) Cryogenic vials and PCR or microcentrifuge tubes
Material Polypropylene
Positions 25.0
Tube Size 0.83mm
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    

22-038-541

 
simport™ scientific
S50025R
Red Case of 10 Each for $43.08

22-038-540

 
simport™ scientific
S50025B
Blue Case of 10 Each for $43.36

22-038-542

 
simport™ scientific
S50025Y
Yellow Case of 10 Each for $43.36
Description & Specifications

Specifications

Autoclavable No
For Use With (Equipment) Cryogenic vials and PCR or microcentrifuge tubes
Material Polypropylene
Positions 25.0
Tube Size 0.83mm
Type Rack, Tube

  • Holds up to 25 polyprolylene or polystyrene tubes of 10 x 75mm or 12 x 75mm dimension
  • Accommodate all types of screw cap microtubes from 0.5 to 2mL
  • Can store 16 PCR or microcentrifuge tubes of 0.2 to 0.5mL after flipping

Handles and anchor pins not included