missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Simport™ Scientific SLIMSETTE™ M511 Tissue Processing/Embedding Cassettes

A new generation of cassettes featuring four square compartments

$457.56 - $457.56

Specifications

Closure Type Integral Lid
Description Tissue Cassette
Material Acetal Polymer
Writing Face Angle 45°
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
23-045-180 Simport™ Scientific
M5114
Green Pack of 1500 Each for $457.56
23-045-178 Simport™ Scientific
M5112
White Pack of 1500 Each for $457.56
23-045-182 Simport™ Scientific
M5116
Blue Pack of 1500 Each for $457.56
Specifications

Description

  • Made of acetal polymer
  • Design has a unique recessed cover
  • Cassettes are thinner, allowing more to be stacked in automatic labeling machines
  • Each cassette has four square compartments measuring 13mm
  • Large labeling area for easy identification
  • Anterior writing area at a 45° angle

Order Info

3 x 500/Cs.

Description & Specifications

Specifications

Integral Lid
Tissue Cassette
Acetal Polymer
45°