Simport™ Scientific SLIMSETTE™ M510 Biopsy Processing/Embedding Cassettes
A new generation of cassettes that are more compact and easier to use
$462.67 - $462.67
Specifications
Closure Type Integral Lid
Description Biopsy Cassette
Material Acetal Polymer
Writing Face Angle 45°
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
23-045-164 Simport™ Scientific
M5102
White Pack of 1500 Each for $462.67
23-045-168 Simport™ Scientific
M5106
Blue Pack of 1500 Each for $462.67
23-045-165 Simport™ Scientific
M5103
Pink Pack of 1500 Each for $462.67
23-045-172 Simport Scientific
M51010
Lilac Pack of 500 Each for $462.67
Specifications
Description
  • Specially designed to hold biopsy specimens during the embedding process
  • Made of acetal polymer
  • Unique recessed cover
  • Cassettes are thinner, allowing more to be stacked in automatic labeling machines
  • One-piece integral lid
  • Large labeling area for easy identification
  • Anterior writing area at a 45° angle
Order Info
3 x 500/Cs.
Description & Specifications
Specifications
Integral Lid
Biopsy Cassette
Acetal Polymer
45°