Simport™ Scientific SLIMSETTE™ M509SL Tissue Processing/Embedding Cassettes in QuickLoad™ Sleeves
A new generation of cassettes, prepackaged for use in automatic cassette labeling instruments
$431.85 - $444.28
Specifications
Closure Type Integral Lid
Description Tissue Cassette
Material Acetal Polymer
Pore Style Slots
Writing Face Angle 45°
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
36-101-9403 Simport™ Scientific
M50910SL
Lilac Case of 75 Each for $431.85
36-101-9405 Simport™ Scientific
M50912SL
Aqua Case of 75 Each for $431.85
36-101-9404 Simport™ Scientific
M50911SL
Orange Case of 75 Each for $431.85
36-101-9406 Simport™ Scientific
M50916SL
Fluorescent Pink Case of 75 Each for $444.28
36-101-9398 Simport™ Scientific
M5095SL
Yellow Case of 75 Each for $431.85
36-101-9397 Simport™ Scientific
M5094SL
Green Case of 75 Each for $431.85
36-101-9396 Simport™ Scientific
M5093SL
Pink Case of 75 Each for $431.85
36-101-9395 Simport™ Scientific
M5092SL
White Case of 75 Each for $431.85
36-101-9399 Simport™ Scientific
M5096SL
Blue Case of 75 Each for $431.85
36-101-9401 Simport™ Scientific
M5098SL
Tan Case of 75 Each for $431.85
36-101-9402 Simport™ Scientific
M5099SL
Gray Case of 75 Each for $431.85
36-101-9400 Simport Scientific
M5097SL
Peach Case of 75 Each for $431.85
Specifications
Description
  • Made of acetal polymer
  • Transparent sleeve allows clear view of cassettes during printing
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • Lid can be opened and closed as often as necessary, and will always relock securely
  • Large labeling area for easy identification
  • Anterior writing area at a 45° angle
Description & Specifications
Specifications
Integral Lid
Acetal Polymer
45°
Tissue Cassette
Slots