Simport™ Scientific SecurTainer™ I Tamper Evident Specimen Container
SecurTainer™ I Tamper Evident Specimen Container for collection, transport and storage of specimens.
Supplier: Simport™ Scientific C57590MA
Description
- Shatter resistant polypropylene containers
- Latex free
- Closure made of polyethylene
- Incorporates unique tamper evident screw cap ensuring safe transport or storage situations
- For transporting specimens
Specifications
Polypropylene | |
90 mL | |
Polyethylene | |
400/Cs. |
Non-sterile | |
Magenta | |
Ribbed Tamper-evident Screw Cap | |
100 Pk. |