Simport™ Scientific Screw Caps
Polyethylene cap for self-standing non sterile transport tubes
$90.28 - $239.46
Specifications
Color White
Material Polyethylene
Product Type Screw Cap
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Includes Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Includes Price Quantity  
22-040-236 Simport™ Scientific
T550WOS
10mL serum-standing non-sterile transport tubes Silicone O-ring seal Pack of 1000 Each for $239.46
22-040-242 Simport™ Scientific
T552W
5, 10 and 12mL serum-standing non-sterile transport tubes Lip seal Pack of 1000 Each for $90.28
22-040-244 Simport™ Scientific
T5527W
7mL serum-standing non-sterile transport tubes Lip seal Pack of 1000 Each for $90.28
22-040-246 Simport™ Scientific
T55230W
30mL serum-standing non-sterile transport tubes Lip seal Pack of 500 Each for $90.72
Specifications
Description
  • Available with silicone O-ring or inner lip seal
  • Inner lip seal ensures leakproof seal
  • Silicone O-ring provides secure seal
  • Silicone O-ring Seal are tested at 13.8 PSI (95kPa)
Description & Specifications
Specifications
White
Polyethylene
Screw Cap