missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Simport™ Scientific S500-80 UniRack™ Universal Tube Racks

Autoclavable and chemicals resistant polypropylene tube holder

Manufacturer:  Simport™ Scientific S50080AS

 View more versions of this product

Catalog No. 05-699-320



Description

Description

  • Autoclavable, chemical-resistant polypropylene
  • One side holds up to 80 polystyrene or polypropylene tubes with diameter of 10–12mm
  • Accept 1 to 5mL cryogenic vials, 1.5 and 2mL microcentrifuge tubes as well as all types of screw cap microtubes from 0.5 to 2mL
  • Flip over to store up to 60 0.2mL PCR or 0.5mL microcentrifuge tubes
  • Can be laterally attached to one another using supplied anchor pins
  • Two removable handles allow units to be safely stacked and moved
  • With frosted area for labeling
  • Transparent lid provides a clear view of the contents (lids sold separately)

Order Info

Dimensions: 9L × 2 1/2W × 1 in.H (23 × 6.7 × 2.7cm).

Specifications

Specifications

Autoclavable
9 x 2.5 x 1 in. (23 x 6.5 x 2.7cm)
1 in.
0.5 to 2 mL, or 0.2 to 0.5 mL Tubes
23 cm
60 or 80
6.5 cm
Assorted
Screw cap microtubes
2.7 cm
9 in.
Polypropylene
2.5 in.
Tube Rack
Documents