missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Simport™ Scientific E-Z LOAD™ Histosette™ II Tissue Cassette Bases

For use with Micro Writer™, Shur/Mark™ and Lamb™ cassette labeling instruments

$449.21 - $656.72

Specifications

Closure Type Integral Lid
Description Tissue Cassette
Material Acetal Polymer
Pore Style Slots
Writing Face Angle 45°
View More Specs

Products 10
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
22-038-230 View Documents Simport™ Scientific
M4822BA
White Case of 2000 Each for $449.21
22-038-232 View Documents Simport™ Scientific
M4824BA
Green Case of 2000 Each for $656.72
22-038-234 View Documents Simport™ Scientific
M4826BA
Blue Case of 2000 Each for $449.21
22-038-233 View Documents Simport™ Scientific
M4825BA
Yellow Case of 2000 Each for $449.21
22-038-231 View Documents Simport™ Scientific
M4823BA
Pink Case of 2000 Each for $449.21
22-038-235 View Documents Simport™ Scientific
M4827BA
Peach Case of 2000 Each for $449.21
22-038-237 View Documents Simport™ Scientific
M4829BA
Gray Case of 2000 Each for $449.21
22-038-239 View Documents Simport™ Scientific
M48211BA
Orange Case of 2000 Each for $449.21
22-038-252 View Documents Simport™ Scientific
M4828BA
Tan Case of 2000 Each for $449.21
22-038-253 View Documents Simport™ Scientific
M48210BA
Lilac Case of 2000 Each for $449.21
Description

Description

  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density acetal polymer
  • Resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • The anterior writing surface is slanted at a 45° angle
  • Available in 11 non-cytotoxic, non-metallic colors

Order Info

  • Stacks of 50 cassettes each; case contains two boxes of 20 stacks
  • Bases and covers must be ordered separately

Specifications

Specifications

Integral Lid
Acetal Polymer
45°
Tissue Cassette
Slots