missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Simport™ Scientific E-Z LOAD™ Histosette™ II Tissue Cassette Bases
For use with Micro Writer™, Shur/Mark™ and Lamb™ cassette labeling instruments
Supplier: Simport™ Scientific M48211BA
Description
- Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
- Molded from a special high-density acetal polymer
- Resistant to the chemical action of histological solvents
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- The anterior writing surface is slanted at a 45° angle
- Available in 11 non-cytotoxic, non-metallic colors
Order Info
- Stacks of 50 cassettes each; case contains two boxes of 20 stacks
- Bases and covers must be ordered separately
Specifications
Slots | |
Integral Lid | |
Acetal Polymer |
45° | |
Orange | |
Tissue Cassette |