Simport™ Scientific DrainRack™ Slide Trays
$50.74 - $54.77
Specifications
Device Holds 200
Dimensions (L x W x H) 9 x 3 x 1 in. (22.8 x 7.62 x 2.5cm)
Material Polystyrene
For Use With (Equipment) Microscope slide
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
22-045-606 Simport™ Scientific
M710100G
Green Case of 10 Each for $50.74
22-045-605 Simport™ Scientific
M710100B
Blue Case of 10 Each for $50.74
22-045-609 Simport™ Scientific
M710100Y
Yellow Case of 10 Each for $50.74
22-045-608 Simport™ Scientific
M710-100W
White Case of 10 Each for $54.77
22-045-607 Simport™ Scientific
M710100P
Pink Case of 10 Each for $50.74
Specifications Description & Specifications
Specifications
200
9 x 3 x 1 in. (22.8 x 7.62 x 2.5cm)
Polystyrene
Microscope slide