Fisherbrand™ Capinsert™ Cap Inserts
Made of polypropylene.
Manufacturer:  Fisherbrand™ T345W
 View more versions of this product
Catalog No. 05-670-4
Description
Description
The Capinsert™ is used to color code a multitude of products such as tubes and vials. It is inserted on top of the closure and has a write-on frosted area for sample identification.
  • Simplifies sample identification
  • Press into place
  • Frosted writing area on top provides space for additional identifying markings
Order Info
Available in 11 colors; assorted packages contain 100 of each color
Specifications
Specifications
Yes
Polypropylene
Micrewtubes microcentrifuge tubes and other products
Used to color code and write on frosted area for sample identification
Polypropylene
White
White
No
Micrewtubes microcentrifuge tubes and other products
Non-sterile
Documents