Simport™ Scientific M480 Disposable Embedding Cassettes
M480 Disposable Embedding Cassettes for holding and identifying tissue samples in processing, embedding and sectioning procedures during in vitro diagnostics.
Supplier: Simport™ Scientific M4802
Description
- Made of acetal polymer and resistant to chemical action of histological solvents
- Fit securely in microtome chuck adapters
- Accepts standard metal lids that keep specimens safe during processing
- Slanted writing surface for easy sample identification
- Available in three colors
Certifications
CE marked
Specifications
White | |
Acetal polymer | |
Embedding Cassette |