Simport™ Scientific Amplitube™ PCR Reaction Strips
Made of polypropylene. Amplitube™ PCR Reaction Strips are manufactured under the most stringent conditions to attain the highest quality standards.
Supplier: Simport™ Scientific T3203N
Description
- Available with either flat (needle pierceable) or dome-topped individually attached hinged caps
- Hinged caps have integral seal that prevents contamination of lid surface
- Ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfers
- Positive sealing fully protects contents from evaporation during thermal cycle
- Strip fits standard well spacing and can also be used with 8-channel hand-held pipettors
- Easily opened and closed with one hand
- Autoclavable
- Choice of colorless and four non-cytotoxic and non-metallic colors
- Packed in tamper-proof resealable bags
Specifications
Natural | |
125 Pk. | |
Non-sterile | |
Domed | |
Tube |
0.2 mL | |
Standard well spacing and can also be used with 8-channel hand-held pipettors | |
Polypropylene | |
Autoclavable | |
8-Strip PCR Reaction Tube |