missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Simple Bunsen Burner
Choice of LPG or natural gas burner.
Manufacturer: Eisco™ CH0087B
Description
- Nickel-plated burner tube with air regulator and cylindrical connector
- Mounted on pressed-steel base
Order Info
Dimensions: burner tube, 4.3H × 0.5 in. dia. (110 × 13mm); base, 3.4 in. dia. (85mm); overall height, 5.9 in. (150mm).
Specifications
Natural Gas | |
Bunsen Burner | |
4.3 in. | |
3.4 in. | |
13 mm |
Aluminum, Brass, Steel | |
Bunsen Burner | |
142 mm | |
85 mm (Base) |