missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Simethicone, USP, 90.5-99%, Spectrum™ Chemical

S1926, 8050-81-5

Supplier:  Spectrum Chemical Mfg Cor S1926200LTBL

 View more versions of this product

Catalog No. 18606248

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Spectrum™ Chemical Simethicone, USP is an antifoaming agent that is used to reduce bloating from excessive gas. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

8050-81-5
Drum
200 L
O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C
238.46
USP
1
C6H18O4Si3
AMTWCFIAVKBGOD-UHFFFAOYSA-N
3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione
90.5 to 99%

Chemical Identifiers

8050-81-5
238.46
3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione
C6H18O4Si3
AMTWCFIAVKBGOD-UHFFFAOYSA-N
O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C
SDS
Documents

Documents

Promotions

Promotions