Thermo Scientific™ Silylation Grade Solvents
Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions.
$282.53 - $762.85
Products 5
Catalog Number Mfr. No. Standard Components Price Quantity  
Catalog Number Mfr. No. Standard Components Price Quantity  
PI20062
View Documents Promotion Details
Thermo Scientific™
TS20062
Acetonitrile
Pack for $349.97
PI20672
View Documents Promotion Details
Thermo Scientific™
TS20672
Dimethylformamide (DMF)
Pack for $762.85
PI20684
View Documents Promotion Details
Thermo Scientific™
TS20684
Dimethylsulfoxide (DMSO)
Pack for $282.53
PI27530
View Documents Promotion Details
Thermo Scientific™
TS27530
Pyridine
Pack for $486.29
PI27860
View Documents Promotion Details
Thermo Scientific™
TS27860
Tetrahydrofuran
Pack for $529.05
Description
Description
Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions. They are purified, dried, and packaged under nitrogen in convenient Thermo Scientific™ Hypo-Vial™ Sample Storage Vials, and are supplied with elastomer septa to enable immediate access to the sample without exposure to moisture and oxygen. Use polar solvents (acetonitrile, dimethylformamide, dimethylsulfoxide, pyridine, tetrahyrdrofuran) to facilitate reactions; nonpolar organic solvents may be used but will not accelerate the rate of reaction.
Recommended Use
Avoid water or alcohol, as TMS reagents react with active hydrogen.
Avoid enolizable ketones.
Use dimethylformamide for steroids and other large molecules.
Use dimethylsulfoxide to prepare TMS derivatives of tertiary alcohols and some compounds with low solubility in other silylation solvents.
Pyridine is an excellent solvent and reaction medium for MS reactions and is an HCI acceptor in reactions involving organochlorosilanes.
Other commonly used solvents include tetrahydrofuran and acetonitrile.
SDS
missing translation for 'documents'
missing translation for 'documents'
For Research Use Only. Not for use in diagnostic procedures.