Please login to your online account to display your discounted pricing

Silver wool, 99.9+% (metals basis), Alfa Aesar™

$192.41 - $192.41

Chemical Identifiers

Chemical Name or Material Silver wool
CAS 7440-22-4
MDL Number MFCD00003397
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4286006

 
alfa aesar
4286006*
Each for $192.41
Description & Specifications

Specifications

Physical Form Approximately 0.05mm Dia Fibers
Quantity 5g
Chemical Name or Material Silver wool
TSCA TSCA
Assay ≥99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00003397
CAS 7440-22-4
Odor Characteristic