missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver wire, 99.999% (metals basis), Alfa Aesar™, Premion™

Form: approx. 32.94g/m

Manufacturer:  Alfa Aesar 11465CC

 View more versions of this product

Catalog No. AA11465CC



Specifications

Specifications

Silver
7440-22-4
99.999% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
Silver
500cm
Silver-colored wire
Ag
algaedyn
[Ag]
Characteristic
23954
Wire, 2mm (0.08 in.) dia.

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

Documents