missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver wire, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 45071G6

 View more versions of this product

Catalog No. AA45071G6



Specifications

Specifications

Silver
Hard
Silver-colored wire
99.95% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
[Ag]
107.868
CHEBI:9141
Silver
10m
7440-22-4
10.5g/cm3
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
Characteristic
silver
23954
Wire, 0.203mm (0.008 in.) dia.

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

SDS Documents