missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver wire, 99.9% (metals basis), Alfa Aesar™

Form: approx. 8.24g/m, Temper: soft

Manufacturer:  Alfa Aesar 11434G1

 View more versions of this product

Catalog No. AA11434G1



Specifications

Specifications

Silver
Annealed
Silver-colored wire
99.9% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
Silver
1m
7440-22-4
10.5g/cm3
Ag
algaedyn
[Ag]
Characteristic
23954
Wire, 1.0mm (0.04 in.) dia.

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

Documents