missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Wire, 0.64mm (0.025 in.) dia., annealed, 99.9% (Metals basis), Alfa Aesar™

Form: Approx. 3.4 g/m; Temper: soft

Manufacturer:  Alfa Aesar 43325G2

 View more versions of this product

Catalog No. AA43325G2



Specifications

Specifications

Silver
Annealed
Silver-colored wire
99.9% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
Silver
2m
7440-22-4
10.5g/cm3
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
Characteristic
23954
Wire, 0.64mm (0.025 in.) dia.

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Documents