missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver wire, 0.1mm (0.004 in.) dia., Premion™, 99.997% (metals basis)

$62.97 - $62.97

Chemical Identifiers

CAS 7440-22-4
MDL Number MFCD00003397
Chemical Name or Material Silver wire, 0.1mm (0.004 in.) dia.
Grade Premion™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11470G2 SDS Alfa Aesar™
11470G2
2m Each for $62.97
AA11470G6 SDS Alfa Aesar™
11470G6
10m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA11470H2 SDS Alfa Aesar™
11470H2
50m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7440-22-4
MFCD00003397
Silver wire, 0.1mm (0.004 in.) dia.
Premion™

Specifications

≈0.08g/m
7440-22-4
MFCD00003397
Silver wire, 0.1mm (0.004 in.) dia.
Premion™
2m
Wire
Characteristic
99.997%, (metals basis)

Safety and Handling

TSCA : TSCA