Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver wire, 0.05mm (0.002 in.) dia., Premion™, 99.99% (metals basis)

$100.50 - $100.50

Chemical Identifiers

Chemical Name or Material Silver wire
CAS 7440-22-4
Name Note 0.05mm (0.002 in.) dia.
Purity Grade Premion™
MDL Number MFCD00003397
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11467G5

  SDS
alfa aesar™
11467G5
5m Each for $100.50

AA11467H2

  SDS
alfa aesar™
11467H2
50m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11467J4

  SDS
alfa aesar™
11467J4
2 x 50m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

TSCA TSCA
Assay Percent Notes (metals basis)
Physical Form Wire
Weight ≈0.02g/m
Quantity 5m
MDL Number MFCD00003397
Assay 99.99%
Odor Characteristic
Name Note 0.05mm (0.002 in.) dia.
Chemical Name or Material Silver wire
Purity Grade Premion™
CAS 7440-22-4