missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver wire, 0.05mm (0.002 in.) dia., annealed, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 44461J4

 View more versions of this product

Catalog No. AA44461J4



Specifications

Specifications

Silver wire
2 x 50m
7440-22-4
Ag
algaedyn
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
99.99%
≈0.02g/m
0.05mm (0.002 in.) dia., Annealed
(metals basis)
MFCD00003397
Insoluble in water.
[Ag]
Characteristic
23954
Wire, ≈0.02g/m

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents