missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver tubing, 6.35mm (0.25in) OD, 5.84mm (0.230in) ID, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 14252BW

 View more versions of this product

Catalog No. AA14252BW



Specifications

Specifications

Silver tubing
50cm
7440-22-4
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
Characteristic
23954
99.9%
≈5.1g/10cm
OD: 6.35mm (0.25 in.) ID: 5.84mm (0.230 in.), Temper: as drawn
(metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents