missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T133110G

 View more versions of this product

Catalog No. T133110G



Specifications

Specifications

Silver Trifluoromethanesulfonate
10g
CAgF3O3S
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
C(F)(F)(F)S(=O)(=O)[O-].[Ag+]
256.931
256.93
≥98.0% (T)
White-Yellow
2923-28-6
MFCD00013226
QRUBYZBWAOOHSV-UHFFFAOYSA-M
silver;trifluoromethanesulfonate
76223
Crystal-Powder at 20°C

Chemical Identifiers

2923-28-6
256.931
QRUBYZBWAOOHSV-UHFFFAOYSA-M
76223
C(F)(F)(F)S(=O)(=O)[O-].[Ag+]
CAgF3O3S
MFCD00013226
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
silver;trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents