Learn More
Silver trifluoromethanesulfonate, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 175201000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Silver trifluoromethanesulfonate | |
99.0% min. | |
99+% | |
CAgF3O3S | |
MFCD00013226 | |
09,302 | |
Solubility in water: soluble. | |
[Ag+].[O-]S(=O)(=O)C(F)(F)F | |
256.93 | |
256.92 |
2923-28-6 | |
Authentic | |
Glass bottle | |
CF3SO3Ag | |
100g | |
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate | |
QRUBYZBWAOOHSV-UHFFFAOYSA-M | |
silver;trifluoromethanesulfonate | |
76223 | |
99+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 220-882-2
RUO â Research Use Only